siRNA Details
Gene Symbol: Clcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133414.1 | NP_596905.1 | 8 | 911 | ||
XM_006233127.1 | XP_006233189.1 | 8 | 950 | |||
XM_006233128.2 | XP_006233190.1 | 8 | 1009 | |||
XM_008761365.1 | XP_008759587.1 | 8 | 1009 | |||
XM_008761366.1 | XP_008759588.1 | 7 | 848 | |||
GenBank mRNA | AB052919.1 | - | 6 | 767 | ||
AB052920.1 | - | 6 | 767 | |||
AB052921.1 | - | 7 | 869 | |||
AB052922.1 | - | 8 | 911 | |||
AB052923.1 | - | 8 | 931 | |||
AF117330.1 | - | 8 | 899 | |||
BC081736.1 | - | 8 | 987 | 7128078 |
