siRNA Details
| Gene Symbol: Zfp483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133422.1 | NP_596913.1 | 6 | 2265 | ||
| GenBank mRNA | AF277901.1 | - | 6 | 2265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp483| Gene Symbol: Zfp483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133422.1 | NP_596913.1 | 6 | 2265 | ||
| GenBank mRNA | AF277901.1 | - | 6 | 2265 | ||