siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133423.1 | NP_596914.1 | 13, 14 | 1726 | ||
| XM_006250805.2 | XP_006250867.1 | 14, 15 | 2078 | |||
| XM_008769987.1 | XP_008768209.1 | 13, 14 | 3093 | |||
| XM_008769989.1 | XP_008768211.1 | 12, 13 | 3039 | |||
| XM_008769990.1 | XP_008768212.1 | 13, 14 | 3094 | |||
| GenBank mRNA | AF144731.1 | - | 13, 14 | 1726 | ||

Ythdc1
Genomic Map