siRNA Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133424.2 | NP_596915.1 | 12 | 1407 | ||
| GenBank mRNA | AF450248.1 | - | 12 | 1408 | ||
| AY096238.1 | - | 12 | 1332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actn3| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133424.2 | NP_596915.1 | 12 | 1407 | ||
| GenBank mRNA | AF450248.1 | - | 12 | 1408 | ||
| AY096238.1 | - | 12 | 1332 | |||