siRNA Details
| Gene Symbol: Tspan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133526.1 | NP_598210.1 | 2 | 321 | ||
| GenBank mRNA | BC061785.1 | - | 2 | 177 | 5623788 | |
| FQ214867.1 | - | 2 | 229 | |||
| FQ216712.1 | - | 2 | 350 | |||
| FQ227464.1 | - | 2 | 338 | |||
| FQ229522.1 | - | 2 | 239 | |||
| Y13275.2 | - | 2 | 321 | |||

Tspan8
Genomic Map