siRNA Details
| Gene Symbol: Pank4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133531.2 | NP_598215.1 | 18, 19 | 2110 | ||
| GenBank mRNA | AF399873.1 | - | 18, 19 | 2110 | ||
| FQ223969.1 | - | 18, 19 | 2117 | |||
| FQ224186.1 | - | 18, 19 | 2119 | |||

Pank4
Genomic Map