siRNA Details
| Gene Symbol: Il13ra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133538.2 | NP_598222.2 | 8 | 978 | ||
| XM_006257427.2 | XP_006257489.1 | 7 | 846 | |||
| XM_006257428.2 | XP_006257490.1 | 7 | 964 | |||
| XM_006257430.2 | XP_006257492.1 | 6 | 777 | |||
| XM_006257431.2 | XP_006257493.1 | 4 | 485 | |||
| XM_008773388.1 | XP_008771610.1 | 7 | 1118 | |||
| XM_008773389.1 | XP_008771611.1 | 8 | 1005 | |||
| XM_008773390.1 | XP_008771612.1 | 4 | 579 | |||
| XM_008773391.1 | XP_008771613.1 | 4 | 1186 | |||
| GenBank mRNA | AF448818.1 | - | 8 | 978 | ||

Il13ra2
Genomic Map