siRNA Details
| Gene Symbol: Pou2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100639.1 | NP_001094109.1 | 4 | 255 | ||
| XM_008769591.1 | XP_008767813.1 | 4 | 2381 | |||
| XM_008769592.1 | XP_008767814.1 | 4 | 516 | |||
| XM_008769593.1 | XP_008767815.1 | 6 | 715 | |||
| XM_008769594.1 | XP_008767816.1 | 5 | 617 | |||
| XM_008769595.1 | XP_008767817.1 | 6 | 650 | |||
| XM_008769596.1 | XP_008767818.1 | 6 | 2825 | |||
| XM_008769597.1 | XP_008767819.1 | 5 | 2727 | |||
| XM_008769598.1 | XP_008767820.1 | 4 | 2626 | |||
| XM_008769599.1 | XP_008767821.1 | 4 | 279 | |||
| GenBank mRNA | DV214198.1 | - | 4 | 255 | ||

Pou2f1
Genomic Map