siRNA Details
Gene Symbol: Pou2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100639.1 | NP_001094109.1 | 12 | 1316 | ||
XM_008769591.1 | XP_008767813.1 | 12 | 3442 | |||
XM_008769592.1 | XP_008767814.1 | 12 | 1577 | |||
XM_008769593.1 | XP_008767815.1 | 14 | 1776 | |||
XM_008769594.1 | XP_008767816.1 | 13 | 1678 | |||
XM_008769595.1 | XP_008767817.1 | 14 | 1711 | |||
XM_008769596.1 | XP_008767818.1 | 14 | 3886 | |||
XM_008769597.1 | XP_008767819.1 | 13 | 3788 | |||
XM_008769598.1 | XP_008767820.1 | 12 | 3687 | |||
XM_008769599.1 | XP_008767821.1 | 12 | 1340 | |||
GenBank mRNA | D12978.1 | - | 4 | 393 | ||
U17013.1 | - | 7 | 906 |
