siRNA Details
| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133545.1 | NP_598229.1 | 13 | 2405 | ||
| GenBank mRNA | U17971.1 | - | 13 | 2405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn21| Gene Symbol: Ptpn21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133545.1 | NP_598229.1 | 13 | 2405 | ||
| GenBank mRNA | U17971.1 | - | 13 | 2405 | ||