siRNA Details
| Gene Symbol: Ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133546.3 | NP_598230.2 | 2 | 1246 | ||
| GenBank mRNA | AF020618.1 | - | 1 | 1423 | ||
| BC072513.1 | - | 2 | 1228 | 7104357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r15a| Gene Symbol: Ppp1r15a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133546.3 | NP_598230.2 | 2 | 1246 | ||
| GenBank mRNA | AF020618.1 | - | 1 | 1423 | ||
| BC072513.1 | - | 2 | 1228 | 7104357 | ||