siRNA Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133548.1 | NP_598232.1 | 12 | 1433 | ||
| XM_006244289.2 | XP_006244351.1 | 12 | 1579 | |||
| XM_006244290.2 | XP_006244352.1 | 12 | 1580 | |||
| XM_006244291.2 | XP_006244353.1 | 12 | 1552 | |||
| GenBank mRNA | AF355102.1 | - | 12 | 1433 | ||
| BC071178.2 | - | 12 | 1496 | 7100147 | ||
| BC087695.1 | - | 12 | 1481 | 7312553 | ||

Hdgfrp2
Genomic Map