siRNA Details
| Gene Symbol: Hdgfrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133548.1 | NP_598232.1 | 14, 15 | 1784 | ||
| XM_006244289.2 | XP_006244351.1 | 14, 15 | 1930 | |||
| XM_006244290.2 | XP_006244352.1 | 14, 15 | 1931 | |||
| XM_006244291.2 | XP_006244353.1 | 14, 15 | 1903 | |||
| GenBank mRNA | AF355102.1 | - | 14, 15 | 1784 | ||
| BC071178.2 | - | 14, 15 | 1847 | 7100147 | ||
| BC087695.1 | - | 14, 15 | 1832 | 7312553 | ||

Hdgfrp2
Genomic Map