siRNA Details
Gene Symbol: Mlxipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133552.1 | NP_598236.1 | 6, 7 | 1113 | ||
GenBank mRNA | AB074517.1 | - | 6, 7 | 1113 | ||
FN432819.1 | - | 6, 7 | 1080 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mlxipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133552.1 | NP_598236.1 | 6, 7 | 1113 | ||
GenBank mRNA | AB074517.1 | - | 6, 7 | 1113 | ||
FN432819.1 | - | 6, 7 | 1080 |