siRNA Details
| Gene Symbol: Mlxipl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133552.1 | NP_598236.1 | 7 | 1399 | ||
| GenBank mRNA | AB074517.1 | - | 7 | 1399 | ||
| FN432819.1 | - | 7 | 1366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlxipl| Gene Symbol: Mlxipl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133552.1 | NP_598236.1 | 7 | 1399 | ||
| GenBank mRNA | AB074517.1 | - | 7 | 1399 | ||
| FN432819.1 | - | 7 | 1366 | |||