siRNA Details
| Gene Symbol: B3galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133553.2 | NP_598237.2 | 1 | 761 | ||
| GenBank mRNA | AB003478.1 | - | 1 | 732 | ||
| BC126083.1 | - | 1 | 761 | 8375818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3galt4| Gene Symbol: B3galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133553.2 | NP_598237.2 | 1 | 761 | ||
| GenBank mRNA | AB003478.1 | - | 1 | 732 | ||
| BC126083.1 | - | 1 | 761 | 8375818 | ||