siRNA Details
| Gene Symbol: Giot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133563.1 | NP_598247.1 | 4 | 702 | ||
| GenBank mRNA | AB047636.1 | - | 4 | 702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Giot1| Gene Symbol: Giot1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133563.1 | NP_598247.1 | 4 | 702 | ||
| GenBank mRNA | AB047636.1 | - | 4 | 702 | ||