siRNA Details
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133564.1 | NP_598248.1 | 6 | 926 | ||
XM_006235674.2 | XP_006235736.1 | 7 | 4769 | |||
GenBank mRNA | AJ302054.1 | - | 6 | 926 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133564.1 | NP_598248.1 | 6 | 926 | ||
XM_006235674.2 | XP_006235736.1 | 7 | 4769 | |||
GenBank mRNA | AJ302054.1 | - | 6 | 926 |