siRNA Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133571.1 | NP_598255.1 | 10 | 1239 | ||
| GenBank mRNA | D16236.1 | - | 10 | 1239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc25a| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133571.1 | NP_598255.1 | 10 | 1239 | ||
| GenBank mRNA | D16236.1 | - | 10 | 1239 | ||