siRNA Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113403.1 | NP_001106874.1 | 1 | 608 | ||
NM_133574.1 | NP_598258.1 | 1 | 608 | |||
GenBank mRNA | AY079189.1 | - | 1 | 590 | ||
U32990.1 | - | 1 | 608 | |||
U76997.1 | - | 1 | 608 |
Gene Symbol: LOC100912445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748706.3 | XP_003748754.1 | 3 | 804 | ||
XM_008758818.1 | XP_008757040.1 | 3 | 804 | |||
XM_008758819.1 | XP_008757041.1 | 3 | 546 |
