siRNA Details
| Gene Symbol: P2ry14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133577.1 | NP_598261.1 | 1 | 742 | ||
| XM_006232396.2 | XP_006232458.1 | 2 | 7800 | |||
| XM_008760984.1 | XP_008759206.1 | 2 | 6063 | |||
| XM_008760985.1 | XP_008759207.1 | 2 | 3642 | |||
| XM_008760986.1 | XP_008759208.1 | 2 | 3623 | |||
| GenBank mRNA | BC083545.1 | - | 2 | 822 | 7191572 | |
| U76206.1 | - | 1 | 742 | |||
More Information
Set Membership: |
|

P2ry14
Genomic Map