siRNA Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270862.1 | NP_001257791.1 | 10 | 700 | ||
| NM_001270863.1 | NP_001257792.1 | 10 | 682 | |||
| NM_001270864.1 | NP_001257793.1 | 9 | 640 | |||
| NM_133583.2 | NP_598267.2 | 9 | 658 | |||
| GenBank mRNA | AF334105.1 | - | 10 | 664 | ||
| AF334106.1 | - | 9 | 622 | |||
| AJ426424.1 | - | 10 | 713 | |||
| AJ426425.1 | - | 9 | 671 | |||
| AJ426426.1 | - | 10 | 676 | |||
| AJ426427.1 | - | 9 | 634 | |||
| FQ216465.1 | - | 2 | 48 | |||

Ndrg2
Genomic Map