siRNA Details
| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133584.1 | NP_598268.1 | 21 | 2932 | ||
| XM_006233260.2 | XP_006233322.1 | 21 | 2860 | |||
| GenBank mRNA | D89093.1 | - | 21 | 2932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde5a| Gene Symbol: Pde5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133584.1 | NP_598268.1 | 21 | 2932 | ||
| XM_006233260.2 | XP_006233322.1 | 21 | 2860 | |||
| GenBank mRNA | D89093.1 | - | 21 | 2932 | ||