siRNA Details
| Gene Symbol: Ces2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133586.2 | NP_598270.1 | 958 | |||
| XM_008760588.1 | XP_008758810.1 | 8 | 927 | |||
| XM_008760589.1 | XP_008758811.1 | 7 | 933 | |||
| GenBank mRNA | AB010635.1 | - | 958 | |||
| AB191005.1 | - | 995 | ||||
| Gene Symbol: LOC100910040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749155.3 | XP_003749203.1 | 7 | 1023 | ||

Ces2c
Genomic Map