siRNA Details
| Gene Symbol: Dscam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133587.1 | NP_598271.1 | 17 | 3529 | ||
| GenBank mRNA | AF334385.1 | - | 17 | 3529 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dscam| Gene Symbol: Dscam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133587.1 | NP_598271.1 | 17 | 3529 | ||
| GenBank mRNA | AF334385.1 | - | 17 | 3529 | ||
Set Membership: |
|