siRNA Details
| Gene Symbol: Cblb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133601.1 | NP_598285.1 | 7 | 959 | ||
| XM_006248258.2 | XP_006248320.1 | 7 | 1033 | |||
| XM_006248259.2 | XP_006248321.1 | 7 | 1116 | |||
| XM_006248260.2 | XP_006248322.1 | 7 | 1032 | |||
| XM_008768626.1 | XP_008766848.1 | 6 | 934 | |||
| XM_008768627.1 | XP_008766849.1 | 7 | 1229 | |||
| GenBank mRNA | AB071283.1 | - | 7 | 959 | ||

Cblb
Genomic Map