siRNA Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133602.1 | NP_598286.1 | 7, 8 | 596 | ||
| XM_006244292.2 | XP_006244354.1 | 7, 8 | 734 | |||
| GenBank mRNA | AF308818.1 | - | 7, 8 | 596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khsrp| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133602.1 | NP_598286.1 | 7, 8 | 596 | ||
| XM_006244292.2 | XP_006244354.1 | 7, 8 | 734 | |||
| GenBank mRNA | AF308818.1 | - | 7, 8 | 596 | ||