siRNA Details
| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133611.2 | NP_598295.2 | 3, 4 | 978 | ||
| GenBank mRNA | AJ315643.1 | - | 3, 4 | 986 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a13| Gene Symbol: Slc2a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133611.2 | NP_598295.2 | 3, 4 | 978 | ||
| GenBank mRNA | AJ315643.1 | - | 3, 4 | 986 | ||