siRNA Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203334.1 | NP_976079.1 | 9 | 1726 | ||
| XM_008759310.1 | XP_008757532.1 | 9 | 2537 | |||
| XM_008759311.1 | XP_008757533.1 | 10 | 1735 | |||
| GenBank mRNA | AY547309.1 | - | 9 | 1618 | ||
| L21672.2 | - | 9 | 1726 | |||

Slc6a5
Genomic Map