siRNA Details
| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760700.1 | XP_008758922.1 | 4 | 340 | ||
| GenBank mRNA | X83579.1 | - | 4 | 186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk7| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760700.1 | XP_008758922.1 | 4 | 340 | ||
| GenBank mRNA | X83579.1 | - | 4 | 186 | ||