siRNA Details
Gene Symbol: Zhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133620.1 | NP_598304.1 | 3 | 1230 | ||
GenBank mRNA | AB072439.1 | - | 3 | 1230 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133620.1 | NP_598304.1 | 3 | 1230 | ||
GenBank mRNA | AB072439.1 | - | 3 | 1230 |