siRNA Details
| Gene Symbol: Epha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134331.1 | NP_599158.1 | 16 | 2934 | ||
| XM_006237962.2 | XP_006238024.1 | 16 | 3080 | |||
| XM_006237963.2 | XP_006238025.1 | 16 | 3077 | |||
| GenBank mRNA | U21954.1 | - | 16 | 2934 | ||

Epha7
Genomic Map