siRNA Details
| Gene Symbol: Kif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244480.1 | XP_006244542.1 | 7 | 846 | ||
| GenBank mRNA | AF035952.2 | - | 7 | 839 | ||
| AY035403.1 | - | 7 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif6| Gene Symbol: Kif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244480.1 | XP_006244542.1 | 7 | 846 | ||
| GenBank mRNA | AF035952.2 | - | 7 | 839 | ||
| AY035403.1 | - | 7 | 718 | |||