siRNA Details
| Gene Symbol: Ctsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134334.2 | NP_599161.2 | 6 | 882 | ||
| GenBank mRNA | BC062032.1 | - | 6 | 882 | 5623627 | |
| FQ228180.1 | - | 2 | 221 | |||
| FQ232703.1 | - | 2 | 221 | |||
| X54467.1 | - | 6 | 827 | |||

Ctsd
Genomic Map