siRNA Details
Gene Symbol: Nlgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134336.2 | NP_599163.2 | 7 | 1942 | ||
XM_006257054.2 | XP_006257116.1 | 6 | 2041 | |||
XM_006257055.2 | XP_006257117.1 | 5 | 1981 | |||
XM_008773251.1 | XP_008771473.1 | 7 | 2101 | |||
GenBank mRNA | U41663.1 | - | 7 | 1942 |
