siRNA Details
| Gene Symbol: Nlgn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134336.2 | NP_599163.2 | 8 | 2123 | ||
| XM_006257054.2 | XP_006257116.1 | 7 | 2222 | |||
| XM_006257055.2 | XP_006257117.1 | 6 | 2162 | |||
| XM_008773251.1 | XP_008771473.1 | 8 | 2282 | |||
| GenBank mRNA | U41663.1 | - | 8 | 2123 | ||

Nlgn3
Genomic Map