siRNA Details
| Gene Symbol: Il12rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191750.1 | NP_001178679.1 | 9 | 1439 | ||
| XM_006236613.2 | XP_006236675.1 | 11 | 1833 | |||
| XM_006236614.2 | XP_006236676.1 | 11 | 1829 | |||
| XM_008762974.1 | XP_008761196.1 | 11 | 1841 | |||
| XM_008762975.1 | XP_008761197.1 | 11 | 1841 | |||

Il12rb2
Genomic Map