siRNA Details
| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182842.1 | NP_878262.1 | 11 | 1366 | ||
| GenBank mRNA | AB101231.1 | - | 11 | 1366 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camk1g| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182842.1 | NP_878262.1 | 11 | 1366 | ||
| GenBank mRNA | AB101231.1 | - | 11 | 1366 | ||