siRNA Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134364.1 | NP_599191.1 | 5 | 746 | ||
| GenBank mRNA | BC099743.1 | - | 5 | 729 | 7937989 | |
| DQ403101.1 | - | 2 | 226 | |||
| FQ214851.1 | - | 2 | 238 | |||
| FQ216116.1 | - | 3 | 326 | |||
| FQ230301.1 | - | 3 | 365 | |||
| FQ231816.1 | - | 1 | 125 | |||
| M19044.1 | - | 4 | 562 | |||
| M57634.1 | - | 2 | 211 | |||

Atp5b
Genomic Map