siRNA Details
| Gene Symbol: Atp5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134364.1 | NP_599191.1 | 5, 6 | 814 | ||
| GenBank mRNA | BC099743.1 | - | 5, 6 | 797 | 7937989 | |
| DQ403101.1 | - | 2, 3 | 294 | |||
| FQ214851.1 | - | 2, 3 | 306 | |||
| FQ216116.1 | - | 3, 4 | 394 | |||
| FQ230301.1 | - | 3, 4 | 433 | |||
| FQ231816.1 | - | 1, 2 | 193 | |||
| M19044.1 | - | 4, 5 | 630 | |||
| M57634.1 | - | 2, 3 | 279 | |||

Atp5b
Genomic Map