siRNA Details
| Gene Symbol: Atp5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134365.2 | NP_599192.1 | 6 | 696 | ||
| GenBank mRNA | BC063808.1 | - | 6 | 637 | 6920021 | |
| CK845764.1 | - | 522 | ||||
| FQ209675.1 | - | 6 | 595 | |||
| FQ212544.1 | - | 6 | 632 | |||
| FQ214353.1 | - | 6 | 658 | |||
| FQ214371.1 | - | 6 | 632 | |||
| FQ214449.1 | - | 6 | 595 | |||
| FQ214672.1 | - | 6 | 631 | |||
| FQ214784.1 | - | 6 | 630 | |||
| FQ215244.1 | - | 6 | 595 | |||
| FQ215436.1 | - | 6 | 595 | |||
| FQ215480.1 | - | 6 | 595 | |||
| FQ215788.1 | - | 6 | 631 | |||
| FQ215823.1 | - | 6 | 648 | |||
| FQ216062.1 | - | 6 | 597 | |||
| FQ216247.1 | - | 6 | 581 | |||
| FQ216597.1 | - | 6 | 587 | |||
| FQ216773.1 | - | 6 | 595 | |||
| FQ216829.1 | - | 6 | 595 | |||
| FQ216922.1 | - | 6 | 595 | |||
| FQ218701.1 | - | 6 | 631 | |||
| FQ219899.1 | - | 6 | 624 | |||
| FQ223683.1 | - | 6 | 621 | |||
| FQ229261.1 | - | 6 | 655 | |||
| FQ229976.1 | - | 6 | 632 | |||
| FQ232356.1 | - | 6 | 632 | |||
| M35052.1 | - | 6 | 615 | |||
| Gene Symbol: LOC100911417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749372.2 | XP_003749420.1 | 691 | |||

Atp5f1
Genomic Map