siRNA Details
| Gene Symbol: Psd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134370.2 | NP_599197.2 | 9 | 2194 | ||
| XM_006231432.2 | XP_006231494.1 | 9 | 2110 | |||
| XM_008760393.1 | XP_008758615.1 | 8 | 2106 | |||
| XM_008760394.1 | XP_008758616.1 | 9 | 2299 | |||
| XM_008760395.1 | XP_008758617.1 | 9 | 2301 | |||
| XM_008760396.1 | XP_008758618.1 | 9 | 2090 | |||
| GenBank mRNA | AB040468.1 | - | 6 | 1185 | ||

Psd
Genomic Map