siRNA Details
| Gene Symbol: Prl8a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134385.2 | NP_599212.1 | 2 | 219 | ||
| XM_006253904.1 | XP_006253966.1 | 2 | 210 | |||
| XM_006253905.1 | XP_006253967.1 | 2 | 213 | |||
| GenBank mRNA | AF239745.1 | - | 2 | 219 | ||
| AF239746.1 | - | 2 | 216 | |||
| AF239747.1 | - | 2 | 219 | |||
| AF239748.1 | - | 2 | 219 | |||
| BC100138.1 | - | 2 | 212 | 7442617 | ||
| BC127474.1 | - | 2 | 210 | 7443385 | ||

Prl8a9
Genomic Map