siRNA Details
| Gene Symbol: Acsbg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134389.1 | NP_599216.1 | 3 | 334 | ||
| XM_008766203.1 | XP_008764425.1 | 4 | 610 | |||
| XM_008766204.1 | XP_008764426.1 | 4 | 4319 | |||
| GenBank mRNA | AF208125.1 | - | 3 | 334 | ||

Acsbg1
Genomic Map