siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 13 | 1593 | ||
| GenBank mRNA | AB046606.1 | - | 6 | 614 | ||
| AF291466.1 | - | 13 | 1809 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata6| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 13 | 1593 | ||
| GenBank mRNA | AB046606.1 | - | 6 | 614 | ||
| AF291466.1 | - | 13 | 1809 | |||