siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 10 | 1162 | ||
| GenBank mRNA | AB046606.1 | - | 3 | 183 | ||
| AF291466.1 | - | 10 | 1378 | |||
| BC126081.1 | - | 10 | 1095 | |||
| BC158581.1 | - | 10 | 1111 | |||

Spata6
Genomic Map