siRNA Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134392.2 | NP_599219.2 | 12 | 1476 | ||
| GenBank mRNA | AB046606.1 | - | 5 | 497 | ||
| AF291466.1 | - | 12 | 1692 | |||
| BC126081.1 | - | 12 | 1409 | |||
| BC158581.1 | - | 12 | 1425 | |||

Spata6
Genomic Map