siRNA Details
| Gene Symbol: Bzw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134402.3 | NP_599229.1 | 3 | 223 | ||
| GenBank mRNA | AF031483.1 | - | 3 | 251 | ||
| BC063149.1 | - | 3 | 227 | 6890706 | ||
| FQ225034.1 | - | 3 | 199 | |||
| FQ228189.1 | - | 3 | 199 | |||

Bzw2
Genomic Map