siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134404.2 | NP_599231.2 | 11 | 1130 | ||
| XM_008769315.1 | XP_008767537.1 | 11 | 1192 | |||
| XM_008769316.1 | XP_008767538.1 | 11 | 1658 | |||
| GenBank mRNA | AF060173.1 | - | 11 | 1287 | ||

Svop
Genomic Map