siRNA Details
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134405.1 | NP_599232.1 | 8 | 1137 | ||
GenBank mRNA | AB023645.1 | - | 8 | 1137 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134405.1 | NP_599232.1 | 8 | 1137 | ||
GenBank mRNA | AB023645.1 | - | 8 | 1137 |